missing translation for 'onlineSavingsMsg'
Learn More
Antimony shot, 1.5-5mm (0.06-0.2in), Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals Product Code.: 11336567

Antimony shot, 1.5-5mm (0.06-0.2in), Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals

Product Code. 11336567
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11336567

Brand: Thermo Scientific Alfa Aesar 000428.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-36-0
Molecular Formula Sb
Molecular Weight (g/mol) 121.76
MDL Number MFCD00011211 MFCD00134030
InChI Key WATWJIUSRGPENY-UHFFFAOYSA-N
Synonym powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
PubChem CID 5354495
ChEBI CHEBI:30304
SMILES [Sb]

Specifications

Color Silver to Gray
Appearance Solid in various forms
Assay Percent Range 99.9999% (Metals basis)
Quantity 25 g
IUPAC Name distibyne
Odor Odorless
Form Shot
Chemical Name or Material Antimony

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ22H049 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number000428.06,000428.14,000428.22
Lot NumberE29S045 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000428.06,000428.14,000428.22
Lot NumberB03R027 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000428.06,000428.14,000428.22
Lot NumberD27S026 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000428.06,000428.14,000428.22
Lot NumberK21Z048 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000428.06,000428.14,000428.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Antimony shot, 1.5-5mm (0.06-0.2in), Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.