missing translation for 'onlineSavingsMsg'
Learn More

ICP Interference Check Standard 5, SPEX CertiPrep™

Product Code. 10750195
Click to view available options
Quantity:
500 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Sb1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 7440-36-0, 87-69-4, 7697-37-2, 7732-18-5
Packaging Triple-leached LDPE bottle
Quantity 500 mL
Certificate of Analysis Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Molecular Formula Sb
MDL Number MFCD00011211 MFCD00134030
InChI Key WATWJIUSRGPENY-UHFFFAOYSA-N
SMILES [Sb]
IUPAC Name antimony
Molecular Weight (g/mol) 121.76
PubChem CID 5354495
ChEBI CHEBI:30304
Grade Assurance
Solvent or Matrix H2O/0.6% Tart. Acid/Tr. HNO3
Chemical Name or Material Interference Check Standard 5
Show More Show Less

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'