missing translation for 'onlineSavingsMsg'
Learn More

Antimony Standard Metal Solution 10,000ppm, for ICP, Solution HCl 1M, Fisher Chemical

Product Code. 10687602 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 10687602

Brand: Fisher Chemical J/8210/05

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Packaging HDPE plastic bottle
Physical Form Liquid
MDL Number 134030
Color Colorless
Quantity 100 mL
compliance-icons
Product Identifier
  • Antimony solution 10 000 ppm
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.