missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony trichloride, 99.5%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
500 g
2 kg
10 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
10kg
2kg
500g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 223°C |
| Melting Point | 73°C |
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 10 kg |
| Assay Percent Range | 99.4% min. (Iodimetry) |
| Linear Formula | SbCl3 |
| Fieser | 08,19 |
| Merck Index | 15, 696 |
| Solubility Information | Solubility in water: 100g/L (25°C). Other solubilities: soluble in diluted hcl,,soluble in chloroform (+/- 22%),soluble in alcohol,benzene,carbon disulfide,,acetone,tartaric acid,ccl4,insoluble in pyridine,quinoline,organic bases |
| Show More |
Safety and Handling
Product Identifier
- Antimony trichloride
- Danger
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P273-Avoid release to the environment.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur