missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony Trichloride, 99+%, Extra Pure, SLR, Fisher Chemical
180.12 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
250g, Plastic powder jar
Specifications
Boiling Point | 223°C |
Melting Point | 73°C |
CAS | 10025-91-9 |
Color | White |
Physical Form | Solid |
Quantity | 250 g |
Molecular Formula | Cl3Sb |
MDL Number | 11212 |
Formula Weight | 228.11g/mol |
Packaging | Plastic powder jar |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS09_Aquatic_Toxicity.png)
Product Identifier
- Antimony trichloride
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'