missing translation for 'onlineSavingsMsg'
Learn More
Antimony trichloride, ACS reagent, Thermo Scientific Chemicals Product Code.: 10616241

Antimony trichloride, ACS reagent, Thermo Scientific Chemicals

Product Code. 10616241
5 g, Glass bottle
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10616241

Brand: Thermo Scientific Acros 401370050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-91-9
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
Synonym antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
PubChem CID 24814
ChEBI CHEBI:74856
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]

Specifications

Boiling Point 223.0°C
Melting Point 73.0°C
Color White
Physical Form Powder, Crystals and/or Chunks
Quantity 5 g
Linear Formula SbCl3
Fieser 08,19
Merck Index 15,696
Solubility Information Solubility in water: 100g/L (25°C). Other solubilities: soluble in diluted hcl,, soluble in chloroform (+/- 22%), soluble in alcohol, benzene, carbon disulfide,, acetone, tartaric acid, ccl4, insoluble in pyridine, quinoline, organic bases
IUPAC Name trichlorostibane
Formula Weight 228.11
Percent Purity ≥99.0% (Iodimetry)
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Antimony trichloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467864 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number401370050,401375000,401370025,401371000
Lot NumberA0463455 Certificate TypeCertificate of Analysis Date03/07/2024 Catalog Number401370050,401375000,401370025,401371000
Lot NumberA0416565 Certificate TypeCertificate of Analysis Date12/02/2024 Catalog Number401370050,401375000,401370025,401371000
Lot NumberA0444924 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number401370050,401375000,401370025,401371000
Lot NumberA0403689 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number401370050,401375000,401370025,401371000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Antimony trichloride
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Antimony trichloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Antimony trichloride, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.