missing translation for 'onlineSavingsMsg'
Learn More
Antimony(V)-chloride, 99%, anhydrous, Thermo Scientific Chemicals Product Code.: 10489900

Antimony(V)-chloride, 99%, anhydrous, Thermo Scientific Chemicals

Product Code. 10489900
50 g, Glass bottle
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10489900

Brand: Thermo Scientific Acros 201210500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-18-9
Molecular Formula Cl5Sb
Molecular Weight (g/mol) 299.02
MDL Number MFCD00011213
InChI Key VMPVEPPRYRXYNP-UHFFFAOYSA-I
Synonym antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid germanShow MoreShow Less
PubChem CID 24294
IUPAC Name pentachloro-$l^{5}-stibane
SMILES Cl[Sb](Cl)(Cl)(Cl)Cl

Specifications

Boiling Point 92.0°C (30.0 mmHg)
Melting Point 2.8°C
Color Colorless to Yellow
Physical Form Liquid
Quantity 50 g
Assay Percent Range 98.5% min
Linear Formula SbCl5
Fieser 01,42; 04,21; 05,18; 06,22; 07,12; 15,14; 17,17
Merck Index 15, 686
Solubility Information Solubility in water: reacts with water forming hcl and antimonyoxide. Other solubilities: soluble in hcl, carbon disulfide, dichloromethane, soluble in an aqueous solution of tartaric acid,, ccl4 and chloroform
Formula Weight 299.01
Percent Purity 99%
Packaging Glass bottle
Specific Gravity 2.36
Density 2.3600g/mL
Chemical Name or Material Antimony(V)-chloride, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464120 Certificate TypeCertificate of Analysis Date01/07/2024 Catalog Number201210010,201210500,201212500
Lot NumberA0460324 Certificate TypeCertificate of Analysis Date29/02/2024 Catalog Number201210010,201210500,201212500
Lot NumberA0425552 Certificate TypeCertificate of Analysis Date21/12/2023 Catalog Number201210010,201210500,201212500
Lot NumberA0430111 Certificate TypeCertificate of Analysis Date14/05/2022 Catalog Number
Lot NumberA0398180 Certificate TypeCertificate of Analysis Date08/07/2020 Catalog Number201210010,201210500,201212500
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Antimony(V)-chloride
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Antimony(V)-chloride, 99%, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.