missing translation for 'onlineSavingsMsg'
Learn More

Antimony(V) fluoride compound with graphite, 1:1 mixture, Thermo Scientific™

Product Code. 15450847
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 15450847

Brand: Thermo Scientific Alfa Aesar L16962.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 59839-60-0
Quantity 25 g
Molecular Formula CF5Sb
MDL Number MFCD00236643
UN Number UN2923
Synonym pentafluorostibine, c, methane; pentafluorostiborane, antimony pentafluoride; flavan, antimony pentafluoride; methane, methane; pentakis fluoranyl-$l^ 5-stibane, methane-pentafluoro-lambda∼5∼-stibane 1/1, antimony v fluoride compound with graphite, 1:1 mixture
InChI Key NRLSWRYERFGPIB-UHFFFAOYSA-I
SMILES [C].F[Sb](F)(F)(F)F
IUPAC Name methane;pentafluoro-$l^{5}-stibane
Molecular Weight (g/mol) 228.76
PubChem CID 45933604
Formula Weight 228.75
Sensitivity Moisture sensitive
Chemical Name or Material Antimony(V) fluoride compound with graphite, 1:1 mixture
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.