missing translation for 'onlineSavingsMsg'
Learn More
Antimony(V) oxide, 30% in H2O, colloidal dispersion, Thermo Scientific™ Product Code.: 11349267

Antimony(V) oxide, 30% in H2O, colloidal dispersion, Thermo Scientific™

Product Code. 11349267
2.5 kg
Click to view available options
Quantity:
500 g
2.5 kg
This item is not returnable. View return policy

Product Code. 11349267

Brand: Thermo Scientific Alfa Aesar 040128.A4

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Flame retardant

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-60-9
Molecular Formula O5Sb2
Molecular Weight (g/mol) 323.515
MDL Number MFCD00011216
InChI Key LJCFOYOSGPHIOO-UHFFFAOYSA-N
Synonym antimony pentoxide, diantimony pentoxide, antimonic oxide, antimony v oxide, diantimony pentaoxide, stibic anhydride, antimony pentaoxide, apox s, antimony oxide sb2o5, nyacol ago 40
PubChem CID 14813
IUPAC Name (dioxo-$l^{5}-stibanyl)oxy-dioxo-$l^{5}-stibane
SMILES O=[Sb](=O)O[Sb](=O)=O

Specifications

Physical Form Particles in Liquid
Quantity 2.5 kg
Formula Weight 323.5
Concentration or Composition (by Analyte or Components) 30% in H2O
Odor Odorless
Chemical Name or Material Antimony(V) oxide, Colloidal dispersion
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Antimony(V) oxide, 30% in H2O, colloidal dispersion, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.