missing translation for 'onlineSavingsMsg'
Learn More

Antimony(V) oxide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog Number 11350839
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
1314-60-9
O5Sb2
323.515
MFCD00011216
LJCFOYOSGPHIOO-UHFFFAOYSA-N
antimony pentoxide, diantimony pentoxide, antimonic oxide, antimony v oxide, diantimony pentaoxide, stibic anhydride, antimony pentaoxide, apox s, antimony oxide sb2o5, nyacol ago 40
14813
(dioxo-$l^{5}-stibanyl)oxy-dioxo-$l^{5}-stibane
O=[Sb](=O)O[Sb](=O)=O
This item is not returnable. View return policy
1314-60-9
O5Sb2
323.515
MFCD00011216
LJCFOYOSGPHIOO-UHFFFAOYSA-N
antimony pentoxide, diantimony pentoxide, antimonic oxide, antimony v oxide, diantimony pentaoxide, stibic anhydride, antimony pentaoxide, apox s, antimony oxide sb2o5, nyacol ago 40
14813
(dioxo-$l^{5}-stibanyl)oxy-dioxo-$l^{5}-stibane
O=[Sb](=O)O[Sb](=O)=O

It is used as flame retardants in acrylonitrile-butadiene-styrene (ABS) polymer, plastics, rubbers, paper and textiles. It finds use as a flocculant in the production of titanium dioxide. It also finds use in ion-exchange resin. It is also employed as catalyst in certain polymerization and oxidation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 380°C (decomposition)
Color Yellow
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Slightly soluble in water. Slowly dissolves in warm HCl and warm KOH. Insoluble in HNO3
Formula Weight 323.5
Odor Odorless
Appearance Yellowish powder
Density 3.78
Chemical Name or Material Antimony(V) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'