Learn More
Antipyrine, 99%, Thermo Scientific Chemicals
£25.13 - £359.93
Chemical Identifiers
CAS | 60-80-0 |
---|---|
Molecular Formula | C11H12N2O |
Molecular Weight (g/mol) | 188.23 |
MDL Number | MFCD00003146 |
InChI Key | VEQOALNAAJBPNY-UHFFFAOYSA-N |
Synonym | antipyrine, phenazone, antipyrin, phenazon, analgesine, anodynine, anodynin, azophen, fenazone, 2,3-dimethyl-1-phenyl-5-pyrazolone |
PubChem CID | 2206 |
ChEBI | CHEBI:31225 |
IUPAC Name | 1,5-dimethyl-2-phenylpyrazol-3-one |
SMILES | CC1=CC(=O)N(N1C)C2=CC=CC=C2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10589800
|
Thermo Scientific Acros
104971000 |
100 g |
£25.13
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10386210
|
Thermo Scientific Acros
104975000 |
500 g |
£65.73
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10647354
|
Thermo Scientific Acros
104970050 |
5 kg | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
60-80-0 | |
188.23 | |
VEQOALNAAJBPNY-UHFFFAOYSA-N | |
2206 | |
1,5-dimethyl-2-phenylpyrazol-3-one |
C11H12N2O | |
MFCD00003146 | |
antipyrine, phenazone, antipyrin, phenazon, analgesine, anodynine, anodynin, azophen, fenazone, 2,3-dimethyl-1-phenyl-5-pyrazolone | |
CHEBI:31225 | |
CC1=CC(=O)N(N1C)C2=CC=CC=C2 |
Specifications
60-80-0 | |
White | |
Authentic | |
Plastic bottle | |
MFCD00003146 | |
15,703 | |
Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol and chloroform,slightly soluble in ether | |
CC1=CC(=O)N(N1C)C2=CC=CC=C2 | |
188.23 | |
CHEBI:31225 | |
99% | |
Antipyrine, 99% |
109°C to 113°C | |
319°C | |
98.5% min. (Iodimetry) | |
C11H12N2O | |
24,27 | |
antipyrine, phenazone, antipyrin, phenazon, analgesine, anodynine, anodynin, azophen, fenazone, 2,3-dimethyl-1-phenyl-5-pyrazolone | |
VEQOALNAAJBPNY-UHFFFAOYSA-N | |
1,5-dimethyl-2-phenylpyrazol-3-one | |
2206 | |
188.23 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye prot
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 200-486-6
missing translation for 'rtecsNumber' : CD2450000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.