Learn More
Invitrogen™ Anza™ 10 DpnI
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Brand:  Invitrogen™ IVGN0108
340.10 GBP valid until 2024-12-31
Use promo code "21978" to get your promotional price.
 View more versions of this product
Product Code. 15505690
  • £358.00 / 1 set
Explore more special offers
Includes: Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer
Description
Description
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• One buffer for all restriction enzymes
• One digestion protocol for all DNA types
• Complete digestion in 15 min
• Overnight digestion without star activity
Certifications
CoA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
200 μL Anza 10 DpnI
500 μL Anza Buffer (10X)
500 μL Anza Red Buffer (10X)
Store at -20°C.
Not CpG Methylation-Sensitive
Anza™ 10X Red Buffer, Anza™ 10X Buffer
37°C
4000 U
Traditional Cloning
Approved for shipment on Wet or Dry Ice
Dpn I
Yes
No
Restriction Enzyme Cloning System
Anza™