missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Anza™ Buffer Set
Click to view available options
Quantity:
1 set
Unit Size:
1 set
Description
Includes
Anza™ 10X Buffer, Anza™ 10X Red Buffer
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
- One buffer for all restriction enzymes
- One digestion protocol for all DNA types
- Complete digestion in 15 min
- Overnight digestion without star activity
Certifications
CoA
Specifications
Specifications
| Chemical Name or Material | Buffer Set |
| For Use With (Application) | Traditional Cloning |
| Product Line | Anza |
| Quantity | 1 set |
| Includes | 2 mL Anza Buffer (10X) 2 mL Anza Red Buffer (10X) Store at -5 to -30°C. |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction