missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Anza™ 11 EcoRI
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Brand: Invitrogen™ IVGN0116
58.19 GBP valid until 2024-12-31
Use promo code "21978" to get your promotional price.
Includes: Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer
Description
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.• One buffer for all restriction enzymes
• One digestion protocol for all DNA types
• Complete digestion in 15 min
• Overnight digestion without star activity
Certifications
CoA
Specifications
400 μL Anza 11 EcoRI 1 mL Anza Buffer (10X) 1 mL Anza Red Buffer (10X) Store at -20°C. |
|
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive | |
Anza™ 10X Red Buffer, Anza™ 10X Buffer | |
37°C | |
8000 U | |
Traditional Cloning |
Approved for shipment on Wet or Dry Ice | |
EcoR I | |
Yes | |
No | |
Restriction Enzyme Cloning System | |
Anza™ |
For Research Use Only. Not for use in diagnostic procedures.