Learn More
Invitrogen™ Anza™ T4 PNK Kit
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Brand:  Invitrogen™ IVGN2304
Product Code. 15461935
  • £152.00 / 1 set
Explore more special offers
Includes: Anza™ T4 PNK Enzyme (Polynucleotide Kinase), Anza™ 10X T4 PNK Buffer,
Description
Description
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
  • Part of the Anza Restriction Enzyme Cloning System
  • Quick DNA phosphorylation in 15 minutes
  • Heat inactivated in 5 minutes
Certifications
CoA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 reactions Anza T4 PNK Enzyme
0.5 mL Anza 10X T4 PNK Buffer
Store at -5 to -30°C.
Polynucleotide Kinase
50 reactions
Anza™
Approved for shipment on Wet or Dry Ice
Anza™ 1X Buffer, Anza™ 10X T4 PNK Buffer
T4 PNK Kit