missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ AOAC Method QuEChERS Clean-up Kit, Pigmented Fruits and Vegetables with Fat, 15 mL Tube

Use Thermo Scientific™ QuEChERS AOAC 2007.01 Method Clean-up Kits to prepare complex samples for LC-MS or GC-MS analysis of residues, toxins, and other contaminants.

Brand:  Thermo Scientific™ S2-15-PF-AOAC-KIT

Product Code. 17349852

  • £414.00 / Pack of 50
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use Thermo Scientific™ QuEChERS Clean-up Kits to prepare complex sample matrices such as fruits and vegetables for LC-MS or GC-MS analysis of pesticide residues, veterinary drug residues, and toxins using the AOAC 2007.01 Method. Our QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) kits contain pre-packaged, ready-weighed salts, sorbents, and buffers designed to streamline your workflow, provide excellent recoveries for a wide variety of analytes, minimize the potential for error, save time and money, and deliver high-quality, reproducible results.

QuEChERS AOAC 2007.01 Method Clean-up Kits

These kits use dispersive solid phase extraction (dSPE) to purify supernatants obtained during extraction. Clean-up kits are available in prefilled 2 mL and 15 mL tube formats for the following matrices: general fruits and vegetables, pigmented fruits and vegetables, fruits and vegetables containing fats and waxes, and all other food types.

Chemicals used in the AOAC 2007.01 Method

  • Acetonitrile: used to perform single-phase extraction on a sample
  • Anhydrous MgSO4: induces phase separation between water content in sample and acetonitrile layer during liquid-liquid partitioning; removes remaining water from sample during dSPE
  • NaOAc: buffers the sample to stabilize pH
  • PSA: primary secondary amine, used to remove polar matrix components, citric co-extractives, and free fatty acids from sample during dispersive solid phase extraction
  • C18: removes fats, sterols, and other non-polar interferences from sample
  • GCB: graphitized carbon black, removes pigment (not recommended for use with planar pesticides)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

QuEChERS AOAC Clean-up Kit; 400 mg PSA, 400 mg C18, 400 mg GCB, 1200 mg Anhydrous MgSO4 prefilled in 15 mL Tubes; 50/Pk.
Salts in Prefilled Tubes
400 mg PSA, 400 mg C18, 400 mg GCB, 1200 mg Anhydrous MgSO4
15 mL
QuEChERS Clean-up Kit
QuEChERS sample preparation for AOAC 2007.01 Method
Pigmented Fruits and Vegetables with Fat
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.