Learn More
AP3S2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15319-1-AP-20UL
 View more versions of this product
Product Code. 16814454
  • £135.00 / 20µL
Explore more special offers
Description
Description
AP3S2 (Adaptor Related Protein Complex 3 Subunit Sigma 2) is a Protein Coding gene. Among its related pathways are Lysosome. Gene Ontology (GO) annotations related to this gene include protein transporter activity. An important paralog of this gene is AP3S1. Part of the AP-3 complex, an adaptor-related complex which is not clathrin-associated. The complex is associated with the Golgi region as well as more peripheral structures. It facilitates the budding of vesicles from the Golgi membrane and may be directly involved in trafficking to lysosomes. In concert with the BLOC-1 complex, AP-3 is required to target cargos into vesicles assembled at cell bodies for delivery into neurites and nerve terminals.
Specifications
Specifications
AP3S2
Polyclonal
Unconjugated
AP3S2
AP 3 complex subunit sigma 2, AP 3 complex subunit sigma 3B, AP3S2, AP3S3, Sigma 3B adaptin, Sigma adaptin 3b, sigma3b, Sigma3B adaptin
Rabbit
Antigen Affinity Chromatography
RUO
10239, 11778, 683402
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P59780, Q8BSZ2
Ap3s2
AP3S2 Fusion Protein Ag7565
20 μL
Primary
Mouse, Rat, Human