missing translation for 'onlineSavingsMsg'
Learn More

Apatite, naturally occurring mineral, grains, approximately 0.06-0.19in, Thermo Scientific Chemicals

For Grain Mounts, XRD and Microprobe Standards, Identification of Unknown Minerals

£22.61 - £271.35

Chemical Identifiers

CAS 1306-05-4
Molecular Formula Ca5FO12P3
Molecular Weight (g/mol) 504.298
MDL Number MFCD00285511
InChI Key VSIIXMUUUJUKCM-UHFFFAOYSA-D
Synonym fluorapatite, apatite, phosphate rock, unii-m4cm1h238j, phospate rock, fluorapatite ca5f po4 3
PubChem CID 10207414
IUPAC Name pentacalcium;fluoride;triphosphate
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[F-].[Ca+2].[Ca+2].[Ca+2].[Ca+2].[Ca+2]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11319128
SDS View Documents
Thermo Scientific Alfa Aesar
042535.03
1 g
£22.61
1g
Please to purchase this item. Need a web account? Register with us today!
11399118
SDS View Documents
Thermo Scientific Alfa Aesar
042535.06
5 g
£65.22
5g
Please to purchase this item. Need a web account? Register with us today!
11309128
SDS View Documents
Thermo Scientific Alfa Aesar
042535.14
25 g
£271.35
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Apatite is one of a few minerals produced and used by biological micro-environmental systems. The primary use of apatite is in the manufacture of fertilizer, it is a source of phosphorus. It is occasionally used as a gemstone. Green and blue varieties in finely divided form, are pigments with excellent covering power.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Apatite, naturally occurring mineral, grains, approximately 0.06-0.19in, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.