Learn More
Fisherbrand™ Apiezon Grease
Lubricate pumps with this grease. Fisherbrand™ Apiezon Grease is designed for use with Fisherbrand™ rotary vane pumps.
Brand:  Fisherbrand™ 800016
 View more versions of this product
Product Code. 12961141
  • £361.30 / 50g
Explore more special offers
Description
Description
  • Apiezon grease
  • For general use in Fisherbrand rotary vane pumps
TRUSTED_SUSTAINABILITY
Specifications
Specifications
L Grease
20°C 7x10-11 torr
50 g
10°C to 30°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Apiezon Grease > Quantity: 50g