missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ApoA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
£121.00 - £352.00
Specifications
Antigen | ApoA2 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17743634
|
Invitrogen
PA5122236 |
20 μL |
£121.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17733634
|
Invitrogen
PA5122235 |
100 μL |
£352.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
At least 9 distinct polymorphic forms of apolipoproteins are known. The apolipoproteins act as stabilizers of the intact lipoprotein particles. Quantitative measurements of HDL, LDL, and VLDL particles of human serum are often used to estimate an individual's relative risk of coronary heart disease. In addition, quantitative immunological measurements of certain apolipoproteins ( especially A-1 and B) have been suggested to be more accurate estimators of coronary heart disease that measurements of lipoprotein particles (especially HDL and LDL). The apolipoproteins are derived from human plasma by density gradient μgtracentrifugation and HPLC.Specifications
ApoA2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P02652 | |
336 | |
Gln24-Gln100 (Accession P02652), with N-terminal His and GST Tag | |
Primary | |
-20°C | |
APOA2 |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Alp-2; APO A2; APOA2; Apoa-2; ApoAII; Apo-AII; ApoA-II; Apolipoprotein; Apolipoprotein A2; Apolipoprotein AII; apolipoprotein A-II; Apolipoprotein A-II(1-76); Hdl-1; ProapoA-II; Proapolipoprotein A-II; Truncated apolipoprotein A-II | |
APOA2 | |
IgG | |
Antigen affinity chromatography | |
Antibody |