missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOD Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10520-1-AP-20UL
This item is not returnable.
View return policy
Description
Apolipoprotein D (APOD) is a component of high density lipoprotein that has no marked similarity to other apolipoprotein sequences. It has a high degree of homology to plasma retinol-binding protein and other members of the alpha 2 microglobulin protein superfamily of carrier proteins, also known as lipocalins. This glycoprotein is closely associated with the enzyme lecithin:cholesterol acyltransferase - an enzyme involved in lipoprotein metabolism.Specifications
APOD | |
Polyclonal | |
Unconjugated | |
APOD | |
Apo D, APOD, apolipoprotein D | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
347 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P05090 | |
APOD | |
APOD Fusion Protein Ag0812 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |