missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOE Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA578803
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL.
Apo E (Apolipoprotein E) plays an important role in the metabolism of lipids in the plasma, and is also is a constituent of various plasma lipoprotein-lipid particles. The protein is synthesized in liver, brain, spleen, and the kidney, and concentrations are high in interstitial fluids, where it plays a role in distributing cholesterol from cells where cholesterol is in excess to cells which require cholesterol. Apo E also plays an important role in the formation of very low density lipoprotein and chylomicrons. The Apo E gene is located on chromosome 19 at position 19q13. 2.Specifications
APOE | |
Polyclonal | |
Unconjugated | |
APOE | |
AD2; AI255918; APOE; apo-E; ApoE4; APOEA; Apolipoprotein; apolipoprotein E; apolipoprotein E3; apolipoprotein E4; Apoprotein; B2G1; BG; LDLCQ5; LPG; MGC1571 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11816, 25728 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5MG BSA and 0.05MG sodium azide | |
P02650, P08226 | |
APOE | |
E.coli-derived mouse Apolipoprotein E recombinant protein (Position: D55-Q294). | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction