Learn More
Applied Biosystems™ TaqMan™ Fast Universal PCR Master Mix (2X), no AmpErase™ UNG
TaqMan™ Fast Universal PCR Master Mix contains all of the components, excluding the template and primers, for superior real-time qPCR.
Brand: Applied Biosystems™ 4352042
181.65 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.03000kg
Description
- Results in 40 minutes for 40 cycles of PCR in a 20 μL reaction volume when using the Applied Biosystems 7900HT and Fast 7500 Fast real-time PCR systems
- The optimized formulation contains AmpliTaq Fast DNA Polymerase, UP, a highly purified DNA polymerase designed to allow instant hot start, minimizing non-specific product formation and allowing room temperature reaction setup
- A proprietary ROX dye serves as a passive internal reference to normalize non-PCR-related fluorescence fluctuations, for superb precision on Applied Biosystems real-time PCR instruments
- Contains everything needed for TaqMan-based qPCR amplification and detection in a single-tube format
- TaqMan Fast Universal PCR Master Mix contains all of the components, excluding the template and primers, for superior real-time qPCR
- Compatible with TaqMan Gene Expression assays
- Store at 2°C to 8°C
Order Info
Guaranteed minimum shelf life is 60 days.
Specifications
TaqMan™ Fast Universal PCR Master Mix, No AmpErase™ UNG Kit contains two tubes of 1.25 mL each sufficient for 250 20-μL PCR reactions. Store at 2-8°C Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
Tube | |
qPCR | |
Fast | |
ROX (Pre-mixed) | |
Fast Universal PCR Master Mix | |
250 Reactions | |
2X | |
250 Reactions | |
20 μL |
Primer-probe | |
High | |
AmpliTaq DNA Polymerase | |
7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode | |
AmpErase™, TaqMan™ | |
Wet Ice | |
Gene Expression, Genotyping, miRNA analysis | |
2 X | |
DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.