missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Fast Universal PCR Master Mix (2X), no AmpErase™ UNG

Product Code. 10311135 Shop All Applied Biosystems Products
Click to view available options
No. of Reactions:
1250 Reactions
250 Reactions
2500 Reactions
500 Reactions
5000 Reactions
Unit Size:
1 set
1250 reactions
50mL
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10311135

Brand: Applied Biosystems™ 4366072

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Requires

Template and primers

TaqMan™ Fast Universal PCR Master Mix contains all of the components, excluding the template and primers, for superior real-time qPCR.

  • Results in 40 minutes for 40 cycles of PCR in a 20 μL reaction volume when using the Applied Biosystems 7900HT and Fast 7500 Fast real-time PCR systems
  • The optimized formulation contains AmpliTaq Fast DNA Polymerase, UP, a highly purified DNA polymerase designed to allow instant hot start, minimizing non-specific product formation and allowing room temperature reaction setup
  • A proprietary ROX dye serves as a passive internal reference to normalize non-PCR-related fluorescence fluctuations, for superb precision on Applied Biosystems real-time PCR instruments
  • Contains everything needed for TaqMan-based qPCR amplification and detection in a single-tube format
  • TaqMan Fast Universal PCR Master Mix contains all of the components, excluding the template and primers, for superior real-time qPCR
  • Compatible with TaqMan Gene Expression assays
  • Store at 2°C to 8°C

Order Info

Guaranteed minimum shelf life is 60 days.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage TaqMan™ Fast Universal PCR Master Mix, No AmpErase™ UNG Kit contains 2 packs of 2 tubes of 1.25 mL each sufficient for 500 20 μL PCR reactions. Store at 2-8°C

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase AmpliTaq DNA Polymerase
Reaction Speed Fast
For Use With (Equipment) 7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode
Passive Reference Dye ROX (Pre-mixed)
Product Line AmpErase, TaqMan
Product Type Fast Universal PCR Master Mix
Quantity 500 reactions
Sufficient For 500 Reactions
For Use With (Application) Gene Expression, Genotyping, miRNA analysis
Fidelity (vs. Taq) 2 X
No. of Reactions 500 Reactions
Sample Type DNA (Genomic), cDNA
Volume 20 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.