missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Fast Universal PCR Master Mix (2X), no AmpErase™ UNG
Shop All Applied Biosystems Products
Click to view available options
No. of Reactions:
1250 Reactions
250 Reactions
2500 Reactions
500 Reactions
5000 Reactions
Unit Size:
1 set
1250 reactions
50mL
Each
Descrizione
Requires
Template and primers
- Results in 40 minutes for 40 cycles of PCR in a 20 μL reaction volume when using the Applied Biosystems 7900HT and Fast 7500 Fast real-time PCR systems
- The optimized formulation contains AmpliTaq Fast DNA Polymerase, UP, a highly purified DNA polymerase designed to allow instant hot start, minimizing non-specific product formation and allowing room temperature reaction setup
- A proprietary ROX dye serves as a passive internal reference to normalize non-PCR-related fluorescence fluctuations, for superb precision on Applied Biosystems real-time PCR instruments
- Contains everything needed for TaqMan-based qPCR amplification and detection in a single-tube format
- TaqMan Fast Universal PCR Master Mix contains all of the components, excluding the template and primers, for superior real-time qPCR
- Compatible with TaqMan Gene Expression assays
- Store at 2°C to 8°C
Order Info
Guaranteed minimum shelf life is 60 days.
Specifica
Specifica
| Concentration | 2X |
| Content And Storage | TaqMan™ Fast Universal PCR Master Mix, No AmpErase™ UNG Kit contains 10 packs of 2 tubes (1.25 ml each), sufficient for 2500 PCR reactions (20 μl each). Store at 2°C to 8°C. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
| Detection Method | Primer-probe |
| Format | Tube |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
| Polymerase | AmpliTaq DNA Polymerase |
| Reaction Speed | Fast |
| For Use With (Equipment) | 7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode |
| Passive Reference Dye | ROX (Pre-mixed) |
| Vedi altri risultati |
For Research Use Only. Not for use in diagnostic procedures.
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto