missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AmpErase™ Uracil N-Glycosylase (UNG)

Product Code. 10638265 Shop All Applied Biosystems Products
Click to view available options
Quantity:
100 units
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10638265

Brand: Applied Biosystems™ N8080096

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

6 x 1.5mL vial each of GeneAmp 10X PCR Buffer II and MgCl2 sufficient for 100 PCR amplifications of 100μL each.

Enzyme removes any uracil incorporated into single- or double-stranded DNA

AmpErase™ Uracil N-Glycosylase (UNG), part of GeneAmp™ PCR Carry-over Prevention Kit, is 26 kDa ultrapure, recombinant enzyme encoded by E. coli uracil N-glycosylase gene, inserted into E. coli host to direct expression of wild type form of enzyme. Enzyme removes any uracil incorporated into single- or double-stranded DNA.

  • Enzymatic method stops PCR carryover contamination, which prevents false positive result
  • No interference with any PCR or real-time PCR application
  • Optimized for use with GeneAmp™ PCR core reagents and GeneAmp™ instrument systems

PCR, PCR and Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains AmpErase™ Uracil N-glycosylase (UNG) (100 units). Sufficient for 100 PCR amplifications, 100 μl each. Store at -20°C.
PCR Method qPCR, Standard PCR
Polymerase DNA Polymerase
Product Line AmpErase
Product Type Uracil N-Glycosylase for PCR reactions
Quantity 100 units
Shipping Condition Dry Ice
Sufficient For 100 PCR Amplifications
For Use With (Application) Standard PCR

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.