missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AmpliTaq™ 360 DNA Polymerase

Product Code. p-4919360 Shop All Applied Biosystems Products
Click to view available options
Quantity:
100 Units
1000 Units
1500 Units
25 x 1000 Units
250 Units
3000 Units
5000 Units
Unit Size:
25000 units
3000 units
5000 units
Each
This item is not returnable. View return policy

Product Code. 10229284

Brand: Applied Biosystems™ 4398808

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

AmpliTaq 360 DNA Polymerase is supplied at 5U/μL with 10X AmpliTaq 360 PCR Buffer, 25mM M2Cl2, and 360 GC Enhancer. Typically, 1.5mL of each of AmpliTaq 360 Buffer, 25mM MgCl2, and 360 GC Enhancer is supplied with each 250U of enzyme. Storage Buffer Enzyme is supplied in 20mM Tris, pH 9.0, 100mM KCl, 0.1mM EDTA, 1mM DTT, 0.5% (w/v) Tween 20, 50% (v/v) glycerol. Source: Recombinant, modified form of the Thermus aquaticus DNA polymerase gene expressed in E. coli. All solutions are prepared in deionized, filter-sterilized water.

The Applied Biosystems AmpliTaq360 DNA Polymerase, when used with the new enhanced AmpliTaq 360 Buffer and the optional 360 GC Enhancer, amplifies a vast range of DNA sequence contexts.

The Applied Biosystems AmpliTaq360 DNA Polymerase, when used with the new enhanced AmpliTaq 360 Buffer and the optional 360 GC Enhancer, amplifies a vast range of DNA sequence contexts. Compared to the original AmpliTaq DNA Polymerase, AmpliTaq 360 DNA Polymerase is purified by an additional proprietary separation process that reduces contaminating bacterial DNA sequences from the enzyme preparation. This ultra-pure enzyme reduces false positive results, amplifies low-level target sequences, and, when combined with the proprietary AmpliTaq 360 Buffer Kit, promotes the amplification of a variety of templates, including those from bacterial and human genomes.

Features

  • Optimized for the broadest range of targets, from everyday to challenging
  • Unmatched sensitivity and yield
  • Market-leading GC enhancer for robust amplification of GC-rich sequences
  • Achieves the highest quality sequencing data

Notes

  • DreamTaq DNA Polymerase is recommended for superior PCR performance.

Order Info

Approved for shipment at Room Temperature or on Wet Ice

TRUSTED_SUSTAINABILITY

Especificaciones

Concentration 5 U/μL
Content And Storage

• 10X AmpliTaq 360 PCR Buffer, 1.5 mL for each 250 U of enzyme
• 25 mM MgCl2, 1.5 mL for each 250 U of enzyme
• 360 GC Enhancer, 1.5 mL for each 250 U of enzyme

Storage Buffer Enzyme is supplied in 20 mM Tris (pH 9.0), 100 mM KCl, 0.1 mM EDTA, 1 mM DTT, 0.5% (w/v), Tween 20, 50% (v/v) glycerol

Source: Recombinant, modified form of the Thermus aquaticus DNA polymerase gene expressed in E. coli

All solutions are prepared in deionized, filter-sterilized water.

When stored at -15°C to -25°C, this product maintains performance at least until the expiration date on the tube label. Before use, thaw frozen components to room temperature and mix well.
Product is guaranteed up to 12 months as indicated by the expiration date.

Format Stand-alone enzyme
GC-Rich PCR Performance Medium
Polymerase AmpliTaq 360 DNA Polymerase
Exonuclease Activity 5' - 3'
Quantity 100 Units
Fidelity (vs. Taq) 1X
Overhang 3'-A
Reaction Format Separate Components
Size (Final Product) 5 kb or less
Starting Material DNA
Mostrar más Mostrar menos

For Research Use Only. Not for use in diagnostic procedures.

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado