missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AmpliTaq Gold™ 360 Master Mix
GREENER_CHOICE

AmpliTaq Gold 360 PCR Master Mix contains everything required for successful PCR amplification in one convenient package–all components are premixed and premeasured.

ÂŁ160.00 - ÂŁ2980.00

Specifications

Concentration 2X
Format Tube
GC-Rich PCR Performance High
Polymerase AmpliTaq Gold 360 DNA Polymerase
Reaction Speed Standard
View More Specs

Includes: AmpliTaq Gold 360 Master Mix is supplied at 2X concentration and the 360 GC Enhancer. Typically, 1.5mL of the 360 GC Enhancer is supplied with each 5mL of master Mix.. Source: Recombinant, modified form of theThermus aquaticusDNA polymerase gene expressed in E. coli. All solutions are prepared in deionized, filter-sterilized water.

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10460085
GREENER_CHOICE
View Documents Promotion Details
Applied Biosystems™
4398876
1 mL
ÂŁ160.00
1mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10289234
GREENER_CHOICE
View Documents Promotion Details
Applied Biosystems™
4398881
5 mL
ONLINE EXCLUSIVE PRICE
ÂŁ385.00
ÂŁ338.65 / 5mL
Save ÂŁ46.35
12% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10219584
GREENER_CHOICE
View Documents Promotion Details
Applied Biosystems™
4398901
10 x 5 mL
ÂŁ2980.00
Pack of 10
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10259814
GREENER_CHOICE
View Documents Promotion Details
Applied Biosystems™
4398886
50 mL
ÂŁ2855.00
50mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description
Show More
Specifications

Specifications

2X
High
Standard
Sustainable packaging
Room Temp or Wet Ice
1X
3'-A
5 kb or less
Tube
AmpliTaq Gold 360 DNA Polymerase
5' - 3'
PCR Master Mix
Cloning, Hot-start PCR
Built-In Hot Start
SuperMix or Master Mix
DNA
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ AmpliTaq Gold™ 360 Master Mix

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.