missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AmpliTaq Gold™ Fast PCR Master Mix

The Applied Biosystems™ AmpliTaq Gold™ Fast PCR Master Mix is a premix, hot start chemistry for end-point PCR analysis.

Brand:  Applied Biosystems™ 4390941

1110.72 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.


 View more versions of this product

Product Code. 10671176

  • £1666.00 / 2500 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price


Includes: This master mix comes in a 250μL tube, enough for 25 reactions.
This item is not returnable. View return policy

Description

Description

The Applied Biosystems™ AmpliTaq Gold™ Fast PCR Master Mix is a premix, hot start chemistry for end-point PCR analysis. Its optimized chemical design enables to shorten PCR running time from a traditional 2 hours to proximal 40 minutes. It generates high quality PCR products that give high quality sequencing results that customers expect from Applied Biosystems™. Optimized for use in sequencing applications, this kit provides specific, high-yield amplicons that can be easily and robustly sequenced. This premixed master mix works with your existing primers to provide sensitive, specific and reproducible results.

  • Fast Results—Speeds the time from sample to results. Amplifies 600bp length gene in approximately 45 minutes
  • Better Sequencing quality—Specific, high-yield amplicons suitable for sequencing applications that yield low peak-under-peak, longer continous read-length and higher QV-30 count
  • Easy to use—The 2X master mix allows for quick experiment setup
  • No primer annealing restrictions

Speed Time to Discovery

AmpliTaq Gold™ Fast PCR Master Mix, UP brings PCR reaction times down to as little as 45 minutes and delivers robust sequencing data on par with standard PCR reagents such as the classic AmpliTaq Gold™ PCR Master Mix. This new master mix has been designed for use with your existing primers and results in specific, high-yield amplicons for easy, high quality sequencing data. Cycle sequencing can now also be performed in as little as 50 minutes.

Quick Setup, High Sensitivity

This pre-mixed hotstart kit allows you to setup your experiment quickly without sacrificing performance. This kit provides reproducible, sensitive results, allowing you to detect a single copy of a gene in 10 ng of DNA.

Robust Fast Sequencing Results

Get the sequencing results you expect from Applied Biosystems™. The AmpliTaq Gold™ Fast PCR Master Mix, UP is optimized for your sequencing reactions. This kit yields specific high-yield amplicons that provide you with clean sequencing reads including low peak-under-peak, longer read-lengths and superior Q30.

Note: See user's manual or package insert for limited label license, and trademark information.

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• AmpliTaq Gold Fast PCR Master Mix, 2 x 12.5 mL bottles

Store at 2°C to 8°C.
High
Fast
Fast PCR Master Mix
Cloning, Hot-start PCR
1X
2500 Reactions
SuperMix or Master Mix
DNA
Bottle
AmpliTaq Gold DNA Polymerase
5' - 3'
Wet Ice
2X
Built-In Hot Start
3'-A
1.5 kb or less
2.5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.