missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Array Card Calibration Install Kit

This kit is used to maintain your ViiA 7 System with TaqMan™ Array Card Block.

Brand:  Applied Biosystems™ 4432250

1923.75 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 15899682

  • £4275.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Reagents to perform a spectral calibration with FAM™, VIC™, and ROX™ dyes, region-of-interest (ROI) and normalization calibration, and to run an RNase P verification card
This item is not returnable. View return policy

Description

Description

It includes reagents to perform a spectral calibration with FAM™, VIC™, and ROX™ dyes, region-of-interest (ROI) and normalization calibration, and to run an RNase P verification card. Calibration and verification should be run at least every six months. Please see your instrument maintenance guide for full instructions on use. The dye formulations for your ViiA™ 7 System will improve results with multiplexing by more accurately representing fluorescent spectra found in your real-time PCR experiments.

  • Dye formulations for the ViiA 7 System will improve results with multiplexing by more accurately representing fluorescent spectra found in real-time PCR experiments
  • Calibration and verification should be run at least every six months; please see instrument maintenance guide for full instructions

Order Info

Store kit at -15° to -25°C.;

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store Reagent at -15°C ± 25°C
Array Card Calibration Install Kit
1 kit
ViiA™ 7 System
Real Time PCR (qPCR)
Spectral Calibration
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.