missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Centri-Sep™ Spin Columns
Fast and efficient purification of nucleic acids
Brand: Invitrogen™ 401763
This item is not returnable.
View return policy
Description
- Centri-Sep™ Spin Columns use Centri-Sep™ gel that provides excellent recovery of DNA fragments larger than 16bp, while removing greater than 98% of salts, NTPs, and other low molecular weight impurities
- Features of Centri-Sep™ columns: Processes samples faster than ultrafiltration columns
- Achieves rapid and efficient separation
- Permits use of custom buffers
- Convenient 20–100μL sample size
- Dry gel format
- Room temperature storage
- Microcentrifuge format
- Designed for performance: The column design is based on the description of gel filtration for the purification of DNA from nick translation reactions by Sambrook, et.al. Each Centri-Sep; unit consists of a specially fritted microfuge tube, dry gel, a wash tube, and a sample collection tube, all specifically designed for this purpose
- Excellent recovery The Centri-Sep gel provides excellent recovery of DNA fragments with sizes greater than 16bp or a 25-mer, while removing >98% of salts, NTPs, and other unwanted low molecular weight impurities
- Versatile The column gel is hydrated with either reagent-grade water or a buffer of your choice, and spun in a microcentrifuge or swinging-bucket centrifuge to remove the intersititial fluid. Your sample is then applied to the column and spun again, leaving you not only with a sample purified through removal of low molecular weight components, but also exchanged into the buffer of your choice
DNA & RNA Purification & Analysis, DNA Extraction, Sequencing Reaction Purification
Order Info
Shipping Condition: Room Temperature
Specifications
Store at room temperature | |
DNA Extraction | |
32 Columns |
Combs, Pre-filled Cartridge, Tube | |
Centri-Sep™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.