missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ FLoid™ Printer Ink/Paper

For use with FLoid Cell Imaging Station-compatible printer

Brand:  Applied Biosystems™ 4472389

Product Code. 12333593

  • £28.42 / Each
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket

Includes: Color ink and paper set. 18 prints per pack.
This item is not returnable. View return policy

Description

Description

Print beautiful, borderless, credit-card- sized photos in vibrant colors. The sticker-based backing allows to paste directly into lab notebook.

  • All-in-one color ink cartridge and labels compatible with the FLoid printer
  • Color ink and label set produces credit-card-sized label
  • Adhesive back can be attached to lab notebook
  • Water-resistant prints last up to 100 years

Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Printer Ink/Paper
Color ink and paper set. 18 prints per pack.

Store dry at room temperature.
Color Ink, Paper Set
Room Temperature
Floid
Floid™ Cell Imaging System
1 pack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers