Learn More
Applied Biosystems™ GeneAmp™ 10X PCR Buffer I, (2 x 75 mL)
Optimized for use with AmpliTaq™ DNA Polymerase for robust PCR amplification
Brand: Applied Biosystems™ 4379876
230.85 GBP valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Code : 50
Additional Details : Weight : 0.50000kg
Includes: Contains 2 x 75mL of GeneAmp™ 10X PCR Buffer I (500mM KCl, 100mM Tris-HCl, pH 8.3; 15mM MgCl2 and 0.01% (w/v) gelatin). Autoclaved.
Description
- Available in several convenient sizes
- Contains 15 mM MgCl2
- Diluted 1:10, provides optimal pH and ionic strength for PCR amplification with AmpliTaq DNA Polymerase
See user's manual or package insert for limited label license and trademark information.
PCR, PCR and Real-Time PCR, Routine PCR
Order Info
Shipping Condition: Wet ice
Specifications
Contains 2 x 75 mL of GeneAmp™ 10X PCR Buffer I (500 mM KCl, 100 mM Tris-HCl, pH 8.3; 15 mM MgCl2 and 0.01% (w/v) gelatin). Autoclaved. Store at -15° to -25°C. |
|
AmpliTaq DNA Polymerase | |
PCR Buffer | |
Standard PCR | |
10 X | |
150 mL |
Primer-probe | |
GeneAmp™ | |
Wet Ice | |
10X | |
Kit |
For Research Use Only. Not for use in diagnostic procedures.