Learn More
Applied Biosystems™ GeneAmp™ 10X PCR Buffer II & MgCl2
Applied Biosystems GeneAmp 10X PCR Buffer II is optimized for use with AmpliTaq DNA Polymerase, resulting in robust PCR amplification.
£33.16 - £244.00
Specifications
Polymerase | AmpliTaq DNA Polymerase |
---|---|
Product Type | PCR Buffer |
For Use With (Application) | Standard PCR |
Concentration | 10X |
Fidelity (vs. Taq) | 1X |
Includes: 2 x 75mL each of GeneAmp™ 10X PCR Buffer II and MgCl2 sufficient for 100 PCR amplifications of 100μL each.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10219774
|
Applied Biosystems™
N8080010 |
1 x 1.5 mL |
£33.16
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10229724
|
Applied Biosystems™
4379878 |
2 x 75 mL |
£244.00
150mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10219164
|
Applied Biosystems™
N8080130 |
6 x 1.5 mL |
£97.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Applied Biosystems™ GeneAmp™ 10X PCR Buffer II is optimized for use with AmpliTaq™ DNA Polymerase, resulting in robust PCR amplification. The provided MgCl2 solution offers versatility for optimizing PCR.
- Optimized for use with AmpliTaq™ DNA Polymerase, offering maximum PCR performance
- Available in several convenient sizes
- Separate MgCl2 solution offers versatility
GeneAmp™ 10X PCR Buffer II, when diluted 1:10, provides the optimal pH and ionic strength for PCR amplification. GeneAmp™ 10X PCR Buffer II is magnesium ion free. A separate MgCl2 solution is provided. When used together, GeneAmp™ 10X PCR Buffer II and MgCl2 solution offer more versatility in optimizing the magnesium ion concentration to achieve optimal PCR amplification with any specific set of primers and DNA template.
Note: See user's manual or package insert for limited label license and trademark information.
Order Info
Shipping Condition: Wet ice
Specifications
AmpliTaq DNA Polymerase | |
Standard PCR | |
1X |
PCR Buffer | |
10X | |
Kit |
For Research Use Only. Not for use in diagnostic procedures.