missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneAmp™ dNTP Blend (2.5 mM ea)

GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial consists of 1 mL of 2.5 mM each of dATP, dCTP, dGTP, and dTTP, yielding a 10 mM blend of dNTPs.

Brand:  Applied Biosystems™ N8080260

22.09 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10381565

  • £36.81 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial consists of 1 mL of 2.5 mM each of dATP, dCTP, dGTP, and dTTP, yielding a 10 mM blend of dNTPs.

Key advantages of GeneAmp dNTPs
• High purity dNTPs are ideal for use in PCR reactions or other applications
• Guaranteed for PCR performance when used with GeneAmp PCR core reagents and GeneAmp instrument systems
• Premixed dNTP blends are available in various concentrations, offering ease of use

TRUSTED_SUSTAINABILITY
Specifications

Specifications

GeneAmp dNTP Blend, 2.5 mM (1 mL)

Store in a constant temperature freezer at –20°C.
GeneAmp™
Dry Ice
10 mM
Liquid
1 mL
Standard PCR
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.