missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneAmp™ dNTP Blend with dUTP (12.5 mM)

GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH.

Brand:  Applied Biosystems™ N8080270

45.12 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 10407124

  • £90.25 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. GeneAmp dNTPs with dUTP can be used to generate dU-containing PCR products in a carry-over prevention strategy in conjunction with AmpErase Uracil N-glycosylase (UNG).

Highlights
• High purity dNTPs are ideal for use in PCR or other applications
• dNTPs with dUTP are optimized for use in carry-over prevention
• Guaranteed PCR performance when used with GeneAmp PCR core reagents and GeneAmp instrument systems
• Premixed dNTP blends are available in various concentrations, offering ease of use

Notes
• GeneAmp dNTP Blend, 12.5 mM with dUTP contains 2.5 mM each of dATP, dCTP, dGTP and 5.0 mM of dUTP

The PCR process is covered by patents owned by Roche Molecular Systems, Inc. and F. Hoffmann-La Roche Ltd.

Order Info

Shipping Condtion: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

GeneAmp dNTP Blend with dUTP, 12.5 mM (1 mL)

Store at –20°C.
GeneAmp™
Dry Ice
12.5 mM
Liquid
1 mL
Standard PCR
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.