missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYBR™ Power SYBR™ Green PCR Master Mix
Offers superior sensitivity and reproducibility, detecting as few as two copies of a target gene over a broad range of template concentrations.
£88.00 - £2855.00
Specifications
Detection Method | SYBR |
---|---|
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | AmpliTaq Gold DNA Polymerase, AmpliTaq Gold LD |
Reaction Speed | Standard |
Includes: SYBR™ Green 1 Dye, AmpliTaq Gold DNA Polymerase LD, dNTPs with dUTP/dTTP blend, Passive Reference 1, and optimized buffer components
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10685795
|
Applied Biosystems™
4368577 |
1 x 1 mL |
£88.00
40 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10658255
|
Applied Biosystems™
4367659 |
1 x 5 mL |
£366.00
200 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10219284
|
Applied Biosystems™
4368706 |
2 x 5 mL |
£648.00
400 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10209284
|
Applied Biosystems™
4368702 |
5 x 5 mL |
£1552.00
1000 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10668265
|
Applied Biosystems™
4368708 |
10 x 5 mL |
£2835.00
2000 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10310115
|
Applied Biosystems™
4367660 |
1 x 50 mL |
£2855.00
Each |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Get superior nucleic acid quantitation with Applied Biosystems™ Power SYBR™ Green Master Mix.
- Everything needed for SYBR™ Green dye-based PCR amplification and detection in a convenient single-tube format
- Power SYBR™ Green PCR Master Mix contains all of the components (excluding the template and primers) for superior SYBR™ Green reagent-based real-time PCR
- The optimized formulation contains highly purified AmpliTaq Gold™ DNA Polymerase, LD to offer increased sensitivity
- Contains a blend of dTTP/dUTP that is compatible with AmpErase™ UNG to minimize carryover contamination
- Includes a proprietary version of ROX™ dye, an internal passive reference, to normalize non-PCR–related fluorescence fluctuations
- This minimizes well-to-well variability that can result from a variety of causes, such as pipetting error and sample evaporation
- Power SYBR™ Green reagent-based PCR chemistry easily replaces the existing SYBR™ Green PCR Master Mix in Applied Biosystems™ protocols using the same setup and thermal cycling conditions
- Store at -20°C
Order Info
Shipping Condition: Wet Ice
Specifications
SYBR | |
qPCR | |
Standard | |
SYBR Green | |
SYBR™ | |
LD (Low DNA) | |
2X |
High | |
AmpliTaq Gold DNA Polymerase, AmpliTaq Gold LD | |
7300 System, 7500 System, 7700 System, 7900HT Fast System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System | |
ROX (Pre-mixed) | |
Real Time PCR SYBR Master Mix | |
Gene Expression | |
DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.