Learn More
Applied Biosystems™ MeltDoctor™ HRM Calibration Plate, Fast 96-well
These plates provide thermal and optical calibration required for accurate and reproducible discrimination between melt curves.
Brand: Applied Biosystems™ 4425618
315.90 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Includes: Contains: 1 plate, 96-well fast format
Description
Applied Biosystem™ ready-to-use MeltDoctor™ HRM Calibration Plates contain all the components required for pure dye and HRM calibration, reducing the complexity of getting started with HRM experimentation.
Ready-to-use MeltDoctor HRM Calibration Plates contain all the components required for pure dye and HRM calibration, reducing the complexity of getting started with HRM experimentation.
Fast Real-Time PCR, Gene Expression Analysis & Genotyping, Genotyping & Genomic Profiling, High Resolution Melting (HRM) Analysis, Methylation Analysis, Methylation-Sensitive HRM Analysis, PCR & Real-Time PCR, RNAi, Epigenetics & Noncoding RNA Research, Real Time PCR (qPCR)
Order Info
Dry ice shipping
Specifications
96-well Plate | |
HRM (High Resolution Melting) Dye | |
Fast | |
HRM Calibration Plate | |
Dry Ice | |
Calibration Plate |
1 plate, 96-well fast format Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
7500 Fast System, 7900HT Fast System | |
MeltDoctor™ | |
Real Time PCR (qPCR) | |
1 x 96-well plate | |
Thermal Calibration, Optic Calibration |
For Research Use Only. Not for use in diagnostic procedures.