Learn More
Applied Biosystems™ MeltDoctor™ HRM Reagent Kit
Provides individually packaged PCR components and the MeltDoctor™ HRM Dye required for high resolution melting analysis.
Brand: Applied Biosystems™ 4425557
464.02 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00450kg
Includes: AmpliTaq Gold™ 360 DNA Polymerase, AmpliTaq Gold 360 Buffer, 360 GC Enhancer, GeneAmp™ dNTP Blend, MeltDoctor HRM Dye (20X)
Description
High Resolution Melting (HRM) analysis is a post-PCR analysis method used to identify genetic variation in nucleic acid sequences.
High Resolution Melting (HRM) analysis is a post-PCR analysis method used to identify genetic variation in nucleic acid sequences.
This kit includes:
- AmpliTaq Gold™ 360 DNA Polymerase which, when combined with AmpliTaq Gold™ 360 Buffer and 360 GC Enhancer, amplifies a vast range of DNA sequence contexts. AmpliTaq Gold™ 360 DNA Polymerase delivers 360° coverage for a full range of targets
- GeneAmp™ dNTP Blend
- MeltDoctorHRM Dye, a stabilized form of the fluorescent SYTO™ 9 double-stranded nucleic acid stain developed by Molecular Probes
Gene Expression Analysis & Genotyping, Genetic Variation Analysis Using Real-Time PCR, Genotyping & Genomic Profiling, High Resolution Melting (HRM) Analysis, Methylation Analysis, Methylation-Sensitive HRM Analysis, PCR & Real-Time PCR, RNAi, Epigenetics & Noncoding RNA Research, Real Time PCR (qPCR)
Order Info
Guaranteed minimum shelf life is 60 days.
Specifications
This kit includes: AmpliTaq Gold™ 360 DNA Polymerase (2 x 50 μL, each at 5 U/μL), AmpliTaq Gold™ 360 Buffer, 360 GC Enhancer, GeneAmp™ dNTP Blend, and MeltDoctor™ HRM Dye (20x). The kit provides sufficient materials for 250 20-μL reactions when using 2 U AmpliTaq Gold 360 DNA Polymerase per reaction. Store at -15° to -25° Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
High | |
AmpliTaq Gold 360 DNA Polymerase | |
7500 Fast System, 7900HT Fast System | |
High Resolution Melting (HRM), SYTO 9 | |
Hrm Reagent Kit | |
DNA (Genomic), cDNA |
HRM | |
qPCR | |
HRM (High Resolution Melting) Analysis, Mutation Scanning | |
SNPs (Unknown or Numerous), Epigenetic Targets | |
MeltDoctor™ | |
Real Time PCR (qPCR) |
For Research Use Only. Not for use in diagnostic procedures.