missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ 12-Cap Strip

Product Code. 10487294 Shop All Applied Biosystems Products
Click to view available options
Color:
Assorted
Quantity:
1000 Strips (5 pk./200)
200 Strips
For Use With (Equipment):
Thermal Cyclers
Unit Size:
Each
Pack of 12
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10487294

Brand: Applied Biosystems™ N8011534

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Designed to fit on MicroAmp reaction tubes, tube strips, and 96-well plates

MicroAmp™ Cap Strips have a domed profile and are suitable for end-point PCR. 12-cap strips available in clear or an assorted set of red, orange, blue, and green.

 

  • Will not interfere with sample reading
  • Provides tight seal to minimize evaporation
  • Easy to apply and remove
Minimizes Evaporation
  • Use MicroAmp Caps Strips to seal tubes and reduce possibility of contamination in sample wells and help ensure reliable results
  • Easy to Use
  • Use MicroAmp Cap Installing Tool (sold separately) to push caps down and form tight seal
  • When ready, just peel away caps using tabs at end
  • Colored for Easy Organization
  • Available in different colors to help track tubes during assay preparation

 

See user's manual or package insert for limited label license and trademark information.

PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified DNA/Rnase Free
Color Assorted
Quantity 1000 Strips (5 pk./200)
For Use With (Equipment) Thermal Cyclers
Product Line MicroAmp™ 
Material Polypropylene
Content And Storage Contains 1,000 MicroAmp™ 12-Cap Strips (5 packs of SKU# N8010534, Qty 200 per pack). Store at room temperature.
Closure Type Domed Cap
No. of Wells 96
Format 1,000 Strip Caps
Product Type 12-Strip Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.