missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ 8-Cap Strip, clear

Designed to fit on MicroAmp reaction tubes, tube strips, and 96-well plates

Brand:  Applied Biosystems™ N8010535

 View more versions of this product

Product Code. 10209104

  • £83.25 / Pack of 300

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

MicroAmp™ Cap Strips have a domed profile and are suitable for end-point PCR. 8-cap strips available in clear or an assorted set of red, orange, blue, and green.

  • Will not interfere with sample reading
  • Provides tight seal to minimize evaporation
  • Easy to apply and remove
Minimizes Evaporation
  • Use MicroAmp Caps Strips to seal tubes and reduce the possibility of contamination in sample wells and help ensure reliable results
  • Easy to Use
    • Use MicroAmp Cap Installing Tool (sold separately) to push caps down and form tight seal
    • When ready, just peel away caps using tabs at end
    Colored for Easy Organization
    • Available in different colors to help track tubes during assay preparation
    • For real-time PCR applications,use MicroAmp Optical 8-Cap Strips (Mfr. No. 4323032) with flat-top profile

    PCR, PCR & Real-Time PCR

    TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    Certified DNA/Rnase Free
    300 Strips
    MicroAmp™ 
    Strip of 8 attached caps for use with 0.2 mL individual tubes, strip tubes (0.1 or 0.2 mL), or 48- or 96-well plates. Natural (clear) color. Store at room temperature.
    Domed Cap
    300 Strip Caps
    Natural
    Thermal Cyclers
    Polypropylene
    Room Temperature
    96
    8-Strip Cap
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents Certificates

    Certificates

    A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

    5 results found
    Lot Number Certificate Type Date Catalog Number
    Lot NumberI08N25Q311 Certificate TypeCertificate of Analysis Date08/04/2025 Catalog Number
    Lot NumberI07N25Q622 Certificate TypeCertificate of Analysis Date08/04/2025 Catalog Number
    Lot NumberI08N25Q111 Certificate TypeCertificate of Analysis Date08/04/2025 Catalog Number
    Lot NumberI07N25Q611 Certificate TypeCertificate of Analysis Date08/04/2025 Catalog Number
    Lot NumberI07N25Q312 Certificate TypeCertificate of Analysis Date08/04/2025 Catalog Number
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Applied Biosystems™ MicroAmp™ 8-Cap Strip, clear > 300 strips

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.