missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ MicroAmp™ 8-Cap Strip, clear
Designed to fit on MicroAmp reaction tubes, tube strips, and 96-well plates
Brand: Applied Biosystems™ N8011535
184.11 GBP valid until 2024-09-27
Use promo code "21650" to get your promotional price.
This item is not returnable.
View return policy
Description
MicroAmp™ Cap Strips have a domed profile and are suitable for end-point PCR. 8-cap strips available in clear or an assorted set of red, orange, blue, and green.
- Will not interfere with sample reading
- Provides tight seal to minimize evaporation
- Easy to apply and remove
- Use MicroAmp Caps Strips to seal tubes and reduce the possibility of contamination in sample wells and help ensure reliable results
- Use MicroAmp Cap Installing Tool (sold separately) to push caps down and form tight seal
- When ready, just peel away caps using tabs at end
- Available in different colors to help track tubes during assay preparation
- For real-time PCR applications,use MicroAmp Optical 8-Cap Strips (Mfr. No. 4323032) with flat-top profile
PCR, PCR & Real-Time PCR
Specifications
Certified DNA/Rnase Free | |
1500 Strips (5 pk./300) | |
MicroAmp™ | |
Includes: 5 packs of N8010535 (Qty 300 per pack) Strip of 8 attached caps for use with 0.2 mL individual tubes, strip tubes (0.1 or 0.2 mL), or 48- or 96-well plates. Natural (clear) color. |
|
96 | |
8-Strip Cap |
Natural | |
Thermal Cyclers | |
Polypropylene | |
Domed Caps | |
1,500 Strip Caps |