missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ Adhesive Film Applicator

Product Code. 10075764 Shop All Applied Biosystems Products
Click to view available options
Quantity:
5 applicators
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10075764

Brand: Applied Biosystems™ 4333183

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Five Adhesive Seal Applicators

Use to form a secure seal between a microplate and an adhesive film

Form a Tight Seal

  • The MicroAmp Adhesive Film Applicator enables you to ergonomically push an adhesive film down upon a microplate to form a tight seal and minimize air bubbles
  • This tight seal will prevent well-to-well contamination and sample loss
Quick and Easy
  • Use the MicroAmp Adhesive Film Applicator to speed the process of sealing multiple microplates
  • This tool is designed according to the microplate dimensions so you can quickly seal a microplate
  • The rugged proprietary plastic endures constant use for large screening applications

See user's manual or package insert for limited label license and trademark information.

PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Line MicroAmp
Certifications/Compliance Certified DNA/RNase Free
Color Natural
Includes Includes: 5 Adhesive Seal Applicators
Material Polyethylene
No. per Pack Pack of 5
Storage Requirements Room Temperature
Product Type Adhesive Film Applicator
Quantity 5 applicators
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.