Learn More
Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 384-Well Clear Reaction Plates with Barcode
Ideal solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments
Brand:  Applied Biosystems™ 4483273
3443.75 GBP valid until 2025-12-31
Use promo code "24286" to get your promotional price.
 View more versions of this product
Product Code. 15252995
  • £3725.00 / Pack of 500
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Validated with Applied Biosystems instruments for optimal fit and performance.
  • Validated with Applied Biosystems instruments for optimal fit and performance
  • Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
  • Constructed to ANSI™/SBS standard, well suited to high-throughput laboratories and robot handling
  • Great mechanical stability and flatness for complete workflow needs
  • Use of polypropylene and high temperature molding process helps prevent DNA, RNA, and enzymes from binding, while polycarbonate provides mechanical stability, flatness, and high durability
  • Ideal choice for use with automation/robotics and multi-instrument experiments (like thermal cycler, real-time thermal cycler, genetic analyzer)
  • Thin-wall construction allows for snug fit to thermal blocks with excellent heat transfer from thermal block to sample
  • Easy-to-read, black well identification text helps to minimize pipetting mistakes and improves sample preparation productivity
  • Platform: 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 7900HT System, GeneAmp™ 9700, QuantStudio™, QuantStudio Dx, Veriti™ Dx Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA 7 System
PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Includes Barcode
Store at room temperature.
MicroAmp
Microplate
Skirted
40 μL
500 Plates
Certified DNA/RNase Free
500 Plates
Optical, Clear
Thermal Cycler, Genetic Analyzer, DNA Analyzer
Polycarbonate, Polypropylene
384
Standard