missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ Optical Adhesive Film

Reduces chance of well-to-well contamination and sample evaporation when applied to a microplate

£82.00 - £235.00

Specifications

Color Optical
Material Polyester
For Use With (Equipment) 2720 Thermal Cycler, 6100 Thermal Cycler, 7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, GeneAmp 9700, QuantStudio™, SimpliAmp™ Thermal Cycler, StepOnePlus™, Veriti Dx Fast Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Fast Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA™ 7 System, ViiA™ 7 Dx Fast System, QuantStudio™ Dx, 7500 Fast Dx System, ViiA™ 7 Fast System, ProFlex™ PCR System
Shipping Condition Room Temperature
Product Type Film, Seal, or Applicator
View More Specs

Includes: 100 Optical Adhesive Covers

Products 2
Product Code Brand No. per Case Price Quantity & Availability  
Product Code Brand No. per Case Price Quantity & Availability  
10299204
View Documents Promotion Details
Applied Biosystems™
4311971
100 Covers
£235.00
Pack of 100
In Stock
to see stock.
Add to basket
 
10567414
View Documents
Applied Biosystems™
4360954
25 Covers
£82.00
Pack of 25
In Stock
to see stock.
Add to basket
 
Description

Description

Applied Biosystems MicroAmp Optical Adhesive Film is designed to reduce well-to-well contamination and sample evaporation. The easy-to-use film is applied by peeling away the backing and firmly pressing the adhesive film across the top of a microplate. This tight seal will help ensure consistent real-time PCR data without obscuring samples during reading, and the protective covering allows microplates to be more easily stacked for space-saving convenience and storage.

Features

  • Pressure-sensitive adhesive provides a tight seal over every well
  • Will not interfere with sample reading
  • Easy-to-apply film will not stick to gloves
  • Prevents sample loss and reduces cross-contamination
  • Designed to fit MicroAmp 96- and 384-well plates

Notes

Thermo Fisher Scientific has performed exhaustive functional testing regarding minor visual inconsistencies (e.g., wrinkles, line impressions, haziness, white spots, etc.) of the MicroAmp Optical Adhesive Film that may result from removal of the liner or application to a plate. These minor visual inconsistencies do not affect the performance of the adhesive tape. Should you have any additional questions, please contact Technical Support.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Optical
2720 Thermal Cycler, 6100 Thermal Cycler, 7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, GeneAmp 9700, QuantStudio™, SimpliAmp™ Thermal Cycler, StepOnePlus™, Veriti Dx Fast Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Fast Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA™ 7 System, ViiA™ 7 Dx Fast System, QuantStudio™ Dx, 7500 Fast Dx System, ViiA™ 7 Fast System, ProFlex™ PCR System
Film, Seal, or Applicator
Pressure Sensitive
Polyester
Room Temperature
MicroAmp
96/384-well Plate
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.