Learn More
Applied Biosystems™ MicroAmp™ Optical Adhesive Film Kit
Designed to fit on MicroAmp Optical 96-well Reaction Plates
Brand: Applied Biosystems™ 4313663
39.14 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00400kg
Includes: MicroAmp Optical Adhesive Covers (quantity 20), Applicator (quantity 1), MicroAmp Optical Cover Compression Pad (quantity 1)
Description
- MicroAmp Optical Adhesive Film reduces the chance of well-to-well contamination and sample evaporation when applied to a microplate
- MicroAmp Optical Film Compression Pads are designed to support a proper thermal seal between the thermal cycler and reaction plates when using adhesive films
- MicroAmp Adhesive Film Applicator is used to form a tight seal between a microplate and an adhesive film
- Pressure-sensitive adhesive provides a tight seal over every well
- Will not interfere with sample reading
- Press-N-Seal: Align properly to the plate, then press to seal
- Easy to apply—does not stick to your glove
Prevent Loss of Precious Samples
- Use this optically clear adhesive film to seal the samples into the wells of a microplate to reduce the possibility of cross-contamination between sample wells and help ensure consistent real-time PCR data
- Easy to Use
- Simply peel the backing away from the adhesive film and apply film firmly across the top of your microplate
- Stack microplates and read samples as you would normally except with less chance of sample contamination
- MicroAmp Optical Film Compression Pads are designed to support a proper thermal seal between the thermal cycler and reaction plates when using adhesive films
- Use the MicroAmp Adhesive Film Applicator to form a tight seal between a microplate and the adhesive film
- The MicroAmp Adhesive Film Applicator enables use to ergonomically push an adhesive film down on a microplate to form a tight seal and minimize air bubbles. This tight seal will prevent well-to-well contamination and sample loss
- Quick and Easy
- Use the MicroAmp Adhesive Film Applicator to speed the process of sealing multiple microplates
- This tool is designed according to the microplate dimensions so microplate can be quickly sealed
- The rugged proprietary plastic endures constant use for large screening applications
See user's manual or package insert for limited label license and trademark information.
PCR and Real-Time PCR, Real Time PCR (qPCR)
Order Info
Shipping Condition: Room Temperature
Specifications
Optical | |
96 | |
2720 Thermal Cycler, 6100 Thermal Cycler, 7000 System, 7900HT System, GeneAmp 9700, QuantStudio™, ViiA™ 7 Dx System, ViiA™ 7 System, QuantStudio™ Dx | |
Film, Seal, or Applicator | |
Pressure Sensitive |
Polyester | |
Includes: MicroAmp™ Optical Adhesive Covers (quantity 20), Applicator (quantity 1), MicroAmp™ Optical Cover Compression Pad (quantity 1) | |
Room Temperature | |
MicroAmp | |
96-well Plate |