missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MUSTANG PURPLE™ Dye Spectral Calibration Plate, 96-well

Used for MUSTANG PURPLE™ spectral calibration of your 7500, ViiA™ 7, QuantStudio™ 6, QuantStudio™ 7, or QuantStudio™ 12K Flex RT-PCR system with a 96-well block.

Brand:  Applied Biosystems™ 4461599

73.15 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 15820423

  • £133.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21632” to get your promotional price

This item is not returnable. View return policy

Description

Description

The dye formulations will help improve multiplexing results by more accurately representing fluorescent spectra that are part of your real-time PCR experiments.

See instrument maintenance guide for full instructions. The dye formulation will help improve multiplexing results by more accurately representing fluorescent spectra that are part of real-time PCR experiments.

Order Info

Dry ice shipping; store in freezer (-5° to -30°C)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in Freezer (-5°C to -30°C).
7500 System, QuantStudio™ 12k Flex, QuantStudio™ 6 Flex, QuantStudio™ 7, ViiA™ 7 System, QuantStudio™ 7, QuantStudio™ 6, QuantStudio™ 12K
Real Time PCR (qPCR)
1 plate
Mustang Purple™
Dye Spectral Calibration Plate
Dry Ice
Spectral Calibration
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.