Learn More
Applied Biosystems™ Protein Thermal Shift™ Dye Kit
This solution significantly reduces the time and cost associated with screening samples relative to currently available methods.
Brand: Applied Biosystems™ 4461146
91.50 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Includes: Protein Thermal Shift™ Buffer, Protein Thermal Shift™ Dye
Description
Applied Biosystems™ Protein Thermal Shift™ kits allow users to employ a dye that binds to exposed hydrophobic residues to monitor the thermal stability of proteins using a real-time PCR instrument in order to identify buffer conditions that stabilize the protein of interest or screen libraries of ligands for compounds that bind the protein of interest.
Save Time and Money with Sample Screening: This solution significantly reduces the time and cost associated with screening samples relative to currently available methods.
Two Kits to Choose From: The Protein Thermal Shift Dye Kit contains the dye and buffer for performing protein thermal shift experiments, enough for up to 1000 reactions. The Protein Thermal Shift Starter Kit contains the Dye Kit plus a control protein and control ligand for test experiments and troubleshooting. The Protein Thermal Shift Starter Kit components are packaged in two boxes, one to be stored at -20°C and the other at 4°C, for maximum flexibility in reagent formulation.
Simplify Analysis with Protein Thermal Shift Software: Our complete Protein Thermal Shift solution includes analysis software which is compatible with run files generated from five of our real-time PCR systems: StepOne™ Real-Time PCR System, StepOnePlus™ Real-Time PCR system, 7500 Fast Real-Time PCR System, and ViiA™7 Real-Time PCR System.
- Screen samples for relative protein thermal stability
- Screen molecules (e.g. small molecule drug candidates, fragment-based libraries, and antibodies) for ligand binding to proteins in a high-throughput manner
- Screen samples for stability changes after protein point mutations or modifications
- Systematically identify suitable/optimal buffer conditions for the measurement of protein-ligand interactions, production of stable protein preparations, protein crystallization, and storage of proteins
- Quickly QC protein preps to ensure protein integrity and/or purity
Order Info
Shipping Conditions: Room Temperature. Shelf Life: Guaranteed minimum shelf life is 60 days.

Specifications
The Protein Thermal Shift™ Dye Kit contains the dye and buffer for performing protein thermal shift experiments, enough for up to 1000 reactions. Store at 18°C to 25°C. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
qPCR | |
7500 Fast System, 7500 System, StepOne™, StepOnePlus™, ViiA™ 7 System | |
Protein Thermal Shift™ | |
Room Temperature | |
Protein Thermal Shift | |
Protein |
Primer-probe | |
Fluorescence Intensity | |
Dye Kit | |
1,000 reactions | |
Not Target-Specific | |
1000 Reactions |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2823926 | Certificate of Analysis | 29/01/2025 | 4461146 |
2811414 | Certificate of Analysis | 29/01/2025 | 4461146 |
2811371 | Certificate of Analysis | 29/01/2025 | 4461146 |
2823923 | Certificate of Analysis | 29/01/2025 | 4461146 |
2823934 | Certificate of Analysis | 29/01/2025 | 4461146 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.