Learn More
Applied Biosystems™ RNase Inhibitor
No DNase or endonuclease activity
Brand: Applied Biosystems™ N8080119
81.65 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : Weight : 0.01000kg
Includes: 100μL at 20 U/μL. Sufficient for 100 reverse transcription reactions, 20μL each. RNase Inhibitor in Storage Buffer: 20mM HEPES-KOH, pH 7.6; 50mM KCl; 8mM DTT; and 50% (v/v) glycerol.
Description
RNase Inhibitor (ribonuclease inhibitor) is a 50 kDa recombinant enzyme used to inhibit RNase activity. It does not contain DNase or endonuclease activity.
- Inhibits RNase activity—preventing degradation of RNA template
- Lacks DNA endonuclease activity—for better product yield
PCR and Real-Time PCR, Reverse Transcription
Order Info
Shipping Condition: Dry ice
Specifications
50 mM | |
Contents: 100 μL at 20 U/μL. Sufficient for 100 reverse transcription reactions, 20 μl each. RNase Inhibitor in Storage Buffer: 20 mM HEPES-KOH, pH 7.6; 50 mM KCl; 8 mM DTT; and 50% (v/v) glycerol. Store at -20°C. | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.