missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ StepOne™ Real-Time PCR System Spectral Calibration Kit (48 well)

Product Code. 10321415 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10321415

Brand: Applied Biosystems™ 4371433

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Contains three 48-well reaction plates - one background plate and two containing dye standards(plates I & II). Plate I contains FAM, JOE and ROX; Plate II contains SYBR™ Green I and VIC.

Easy-to-use kit establishes the pure dye spectra and multicomponent values for FAM™/SYBR™ Green I, VIC™/JOE™, and ROX™ dyes on the StepOne™ Real-Time PCR System.

The Applied Biosystems™ StepOne™ Real-Time PCR System Spectral Calibration Kit contains one background plate and three preloaded, 48-well fast optical reaction plates containing five dye standards.

  • Easy-to-use
  • Provides accurate discrimination of the fluorescence signal of individual dyes in the reaction mixture
  • Includes the Passive Reference I dye standard for signal normalization in all real-time and endpoint reactions

Order Info

Dry ice shipping; store at -15° to 25°C; protect from light; expiration date: 12 months from the date of delivery.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains three 48-well reaction plates—one background plate and two containing dye standards(plates I & II). Plate I contains FAM, JOE and ROX; Plate II contains SYBR™ Green I and VIC.

Store at -15 to 25°C. Protect from light.

Expiration date: 12 months from the date of delivery
Calibrated Dye FAM, JOE, ROX, VIC, SYBR™ Green
For Use With (Equipment) StepOne™, Fast Mode, StepOne™, Standard Mode
Detection Method Fluorescence
Product Line StepOne
Product Type Spectral Calibration Kit
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Dry Ice
Quantity 1 kit
Calibration Type Spectral Calibration

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.